ENA413172936 Enamine

2-[(2-chloro-6-fluorophenyl)methyl]pyrrolidine

  • MDL number MFCD10691061
  • Cas Number : 1092277-10-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-42822

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-42822
Availability details for
Quantity
  • Availability
  • Add to Cart