ENA413172980 Enamine

2-chloro-N-[(3,5-dimethyl-1-phenyl-1H-pyrazol-4-yl)methyl]acetamide

  • MDL number MFCD26936306
  • Cas Number : 1803582-14-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-46375

Safety Information

msds https://www.enaminestore.com/catalog/EN300-46375
Availability details for
Quantity
  • Availability
  • Add to Cart