ENA413172981 Enamine

4-{[(propan-2-yl)amino]methyl}benzonitrile

  • MDL number MFCD09807206
  • Cas Number : 91350-01-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-32125

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-32125
Availability details for
Quantity
  • Availability
  • Add to Cart