ENA413172987 Enamine

3-phenyl-4,5-dihydro-1H-1,2,4-triazol-5-one

  • MDL number MFCD00457971
  • Cas Number : 939-07-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 329-330
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-49288

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-49288
Availability details for
Quantity
  • Availability
  • Add to Cart