ENA413173009 Enamine

2-(3-methyl-2-oxo-2,3-dihydro-1,3-benzoxazol-5-yl)acetonitrile

  • MDL number MFCD21984798
  • Cas Number : 245095-52-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-50112

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-50112
Availability details for
Quantity
  • Availability
  • Add to Cart