ENA413173023 Enamine

1-amino-2-methylcyclohexane-1-carboxylic acid

  • MDL number MFCD01734664
  • Cas Number : 13725-01-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-51290

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-51290
Availability details for
Quantity
  • Availability
  • Add to Cart