ENA413173053 Enamine

(diphenylmethyl)(ethyl)amine

  • MDL number MFCD09935826
  • Cas Number : 53693-47-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-32314

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-32314
Availability details for
Quantity
  • Availability
  • Add to Cart