ENA413173054 Enamine

ethyl 5-formyl-1H-pyrazole-3-carboxylate

  • MDL number MFCD16620106
  • Cas Number : 93290-12-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-55217

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-55217
Availability details for
Quantity
  • Availability
  • Add to Cart