ENA413173082 Enamine

N,3,4-trimethylaniline

  • MDL number MFCD11152839
  • Cas Number : 38036-47-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-57333

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-57333
Availability details for
Quantity
  • Availability
  • Add to Cart