ENA413173093 Enamine

1-N-(pyridin-4-yl)benzene-1,3-diamine

  • MDL number MFCD09935428
  • Cas Number : 109871-19-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-57352

Safety Information

msds https://www.enaminestore.com/catalog/EN300-57352
Availability details for
Quantity
  • Availability
  • Add to Cart