ENA413173110 Enamine

2-phenylbutan-1-amine

  • MDL number MFCD06212588
  • Cas Number : 34577-88-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-57392

Safety Information

msds https://www.enaminestore.com/catalog/EN300-57392
Availability details for
Quantity
  • Availability
  • Add to Cart