ENA413173198 Enamine

1-(2-aminoethoxy)-2,4-dimethylbenzene

  • MDL number MFCD05855331
  • Cas Number : 50912-65-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-57622

Safety Information

msds https://www.enaminestore.com/catalog/EN300-57622
Availability details for
Quantity
  • Availability
  • Add to Cart