ENA413173205 Enamine

4-[(cyclopropylamino)methyl]-N-methylbenzamide

  • MDL number MFCD14586528
  • Cas Number : 1171310-16-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-57641

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-57641
Availability details for
Quantity
  • Availability
  • Add to Cart