ENA413173292 Enamine

2-fluoro-4-phenylaniline

  • MDL number MFCD06802824
  • Cas Number : 76302-56-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95
Storage temp. Normal

Safety Information

msds https://www.enaminestore.com/catalog/EN300-57801
Availability details for
Quantity
  • Availability
  • Add to Cart