ENA413173545 Enamine

(3-propoxyphenyl)methanamine

  • MDL number MFCD07754795
  • Cas Number : 37806-33-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-58166

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-58166
Availability details for
Quantity
  • Availability
  • Add to Cart