ENA413173551 Enamine

2-methyl-1-(piperidin-4-yl)-2,3-dihydro-1H-indole

  • MDL number MFCD09946214
  • Cas Number : 1016847-63-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-58172

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-58172
Availability details for
Quantity
  • Availability
  • Add to Cart