ENA413173600 Enamine

1-(morpholine-4-carbonyl)cyclohexan-1-amine

  • MDL number MFCD09937079
  • Cas Number : 1016857-95-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-58246

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-58246
Availability details for
Quantity
  • Availability
  • Add to Cart