ENA413173620 Enamine

1,3-diphenylpropan-1-amine

  • MDL number MFCD09813589
  • Cas Number : 5689-74-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-58283

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-58283
Availability details for
Quantity
  • Availability
  • Add to Cart