ENA413173666 Enamine

1-benzyl-1H-imidazol-2-amine

  • MDL number MFCD03412452
  • Cas Number : 22944-67-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 139-140
Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-58361

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-58361
Availability details for
Quantity
  • Availability
  • Add to Cart