ENA413173668 Enamine

4-(pyrrolidin-1-yl)pyridin-3-amine

  • MDL number MFCD10690637
  • Cas Number : 90648-19-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-58364

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-58364
Availability details for
Quantity
  • Availability
  • Add to Cart