ENA413173670 Enamine

methyl[(1-phenyl-1H-pyrazol-3-yl)methyl]amine

  • MDL number MFCD13709541
  • Cas Number : 1156391-85-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-58374

Safety Information

msds https://www.enaminestore.com/catalog/EN300-58374
Availability details for
Quantity
  • Availability
  • Add to Cart