ENA413173770 Enamine

2-[4-(2-aminoethyl)phenoxy]ethan-1-ol

  • MDL number MFCD08449030
  • Cas Number : 4960-64-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-58533

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-58533
Availability details for
Quantity
  • Availability
  • Add to Cart