ENA413173798 Enamine

4-(dimethylamino)-3-fluorobenzoic acid

  • MDL number MFCD11156555
  • Cas Number : 160658-70-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 170-172
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-58580

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-58580
Availability details for
Quantity
  • Availability
  • Add to Cart