ENA413173839 Enamine

1-(4-methoxyphenyl)-2-(morpholin-4-yl)ethan-1-amine

  • MDL number MFCD09262886
  • Cas Number : 927997-72-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-58663

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-58663
Availability details for
Quantity
  • Availability
  • Add to Cart