ENA413173944 Enamine

cyclohexylmethanesulfonamide

  • MDL number MFCD12783435
  • Cas Number : 4352-59-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-59804

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-59804
Availability details for
Quantity
  • Availability
  • Add to Cart