ENA413173951 Enamine

3-chloro-5H,6H,7H-cyclopenta[c]pyridazine

  • MDL number MFCD16683700
  • Cas Number : 872292-64-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95
Storage temp. 0

Safety Information

msds https://www.enaminestore.com/catalog/EN300-60239
Availability details for
Quantity
  • Availability
  • Add to Cart