ENA413173966 Enamine

1-(2-aminoethyl)azepan-2-one

  • MDL number MFCD09808169
  • Cas Number : 27431-27-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-57954

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-57954
Availability details for
Quantity
  • Availability
  • Add to Cart