ENA413173986 Enamine

2-methylpropyl piperazine-1-carboxylate

  • MDL number MFCD11128916
  • Cas Number : 23672-96-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-61642

Safety Information

msds https://www.enaminestore.com/catalog/EN300-61642
Availability details for
Quantity
  • Availability
  • Add to Cart