ENA413173988 Enamine

3-(1,3-thiazol-2-yl)propan-1-amine

  • MDL number MFCD09891571
  • Cas Number : 63779-34-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-61666

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-61666
Availability details for
Quantity
  • Availability
  • Add to Cart