ENA413173994 Enamine

tert-butyl 2-(3-aminopropyl)pyrrolidine-1-carboxylate

  • MDL number MFCD06656818
  • Cas Number : 887587-47-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-62213

Safety Information

msds https://www.enaminestore.com/catalog/EN300-62213
Availability details for
Quantity
  • Availability
  • Add to Cart