ENA413174008 Enamine

2-(4-chlorophenyl)-2H-1,2,3-triazole-4-carbaldehyde

  • MDL number MFCD11169406
  • Cas Number : 41886-24-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 117...119
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-62765

Safety Information

msds https://www.enaminestore.com/catalog/EN300-62765
Availability details for
Quantity
  • Availability
  • Add to Cart