ENA413174012 Enamine

3-(1-methyl-1H-pyrazol-4-yl)piperidine

  • MDL number MFCD20306507
  • Cas Number : 1340528-88-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-62902

Safety Information

msds https://www.enaminestore.com/catalog/EN300-62902
Availability details for
Quantity
  • Availability
  • Add to Cart