ENA413174043 Enamine

2-aminopentan-3-ol

  • MDL number MFCD20232890
  • Cas Number : 116836-16-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-64821

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-64821
Availability details for
Quantity
  • Availability
  • Add to Cart