ENA413174112 Enamine

2-(2-chloropropanamido)-5-phenylthiophene-3-carboxamide

  • MDL number MFCD26936308
  • Cas Number : 1607265-60-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-68515

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-68515
Availability details for
Quantity
  • Availability
  • Add to Cart