ENA413174126 Enamine

4-(4-ethyl-4H-1,2,4-triazol-3-yl)piperidine

  • MDL number MFCD16683621
  • Cas Number : 1249274-35-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-69150

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-69150
Availability details for
Quantity
  • Availability
  • Add to Cart