ENA413174138 Enamine

tert-butyl 3-cyano-3-hydroxyazetidine-1-carboxylate

  • MDL number MFCD21644405
  • Cas Number : 1493737-08-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-69915

Safety Information

msds https://www.enaminestore.com/catalog/EN300-69915
Availability details for
Quantity
  • Availability
  • Add to Cart