ENA413174140 Enamine

2,3-dihydro-1H-indole-5-carboxamide

  • MDL number MFCD11103275
  • Cas Number : 1082311-02-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-70048

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-70048
Availability details for
Quantity
  • Availability
  • Add to Cart