ENA413174170 Enamine

2-(2-methylpiperazin-1-yl)-1,3-benzoxazole

  • MDL number MFCD16842683
  • Cas Number : 207746-95-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-70922

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-70922
Availability details for
Quantity
  • Availability
  • Add to Cart