ENA413174173 Enamine

2-[4-(methylamino)piperidin-1-yl]ethan-1-ol

  • MDL number MFCD12154913
  • Cas Number : 1183836-67-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-71025

Safety Information

msds https://www.enaminestore.com/catalog/EN300-71025
Availability details for
Quantity
  • Availability
  • Add to Cart