ENA413174177 Enamine

1-[methyl(piperidin-4-yl)amino]propan-2-ol

  • MDL number MFCD12764454
  • Cas Number : 1179290-05-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-71088

Safety Information

msds https://www.enaminestore.com/catalog/EN300-71088
Availability details for
Quantity
  • Availability
  • Add to Cart