ENA413174189 Enamine

methyl 2-amino-5-(propan-2-yloxy)benzoate

  • MDL number MFCD14621949
  • Cas Number : 458537-97-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-71502

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-71502
Availability details for
Quantity
  • Availability
  • Add to Cart