ENA413174213 Enamine

4-amino-1,1,1-trifluorobutan-2-ol

  • MDL number MFCD12047592
  • Cas Number : 57165-84-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 50-55
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-73252

Safety Information

signalword Danger
pcodes P210,P264,P270,P280,P301+P312,P305+P351+P338,P310,P330,P370+P378,P403+P235,P501
MSDS https://www.enaminestore.com/catalog/EN300-73252
pictograms GHS05,GHS07
hstatements H227,H302,H318
Availability details for
Quantity
  • Availability
  • Add to Cart