ENA413174216 Enamine

2-chloro-5-ethoxypyridine

  • MDL number MFCD09835252
  • Cas Number : 856851-48-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-73350

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-73350
Availability details for
Quantity
  • Availability
  • Add to Cart