ENA413174248 Enamine

2,5,6,7-tetrahydro-1H-azepin-2-one

  • MDL number MFCD19216497
  • Cas Number : 2228-79-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -20
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-74672

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-74672
Availability details for
Quantity
  • Availability
  • Add to Cart