ENA413174253 Enamine

3-(1H-1,3-benzodiazol-2-yl)-2,3-dihydroxypropanoic acid

  • MDL number MFCD00458799
  • Cas Number : 49671-84-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-74859

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-74859
Availability details for
Quantity
  • Availability
  • Add to Cart