ENA413174294 Enamine

3-amino-1-cyclopropylpyrrolidin-2-one

  • MDL number MFCD14689594
  • Cas Number : 1251066-16-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-77438

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-77438
Availability details for
Quantity
  • Availability
  • Add to Cart