ENA413174326 Enamine

2-chloro-6-(propan-2-yloxy)pyrazine

  • MDL number MFCD09808340
  • Cas Number : 1016698-79-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-79783

Safety Information

msds https://www.enaminestore.com/catalog/EN300-79783
Availability details for
Quantity
  • Availability
  • Add to Cart