ENA413174341 Enamine

1-methyl-1H,4H,5H,6H,7H-[1,2,3]triazolo[4,5-c]pyridine

  • MDL number MFCD19224836
  • Cas Number : 160752-39-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-80813

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-80813
Availability details for
Quantity
  • Availability
  • Add to Cart