ENA413174347 Enamine

2-{ethyl[(9H-fluoren-9-ylmethoxy)carbonyl]amino}acetic acid

  • MDL number MFCD18328448
  • Cas Number : 162545-29-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-81228

Safety Information

msds https://www.enaminestore.com/catalog/EN300-81228
Availability details for
Quantity
  • Availability
  • Add to Cart