ENA413174355 Enamine

1-(propan-2-yl)-4,5,6,7-tetrahydro-1H-indazol-6-amine

  • MDL number MFCD26694114
  • Cas Number : 1334203-56-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-81868

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-81868
Availability details for
Quantity
  • Availability
  • Add to Cart